
Ferrocenium tetrafluoroborate

Empirical Formula (Hill Notation):
C10H10BF4Fe
CAS Number:
Molecular Weight:
272.84
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Reagent Type
Core
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing