Skip to Content
Merck
vibrant-m

Febantel Related Compound B

vibrant-m
Synonym(s):
2-(Methoxymethyl)-5-(phenylthio)-1H-benzimidazole
Empirical Formula (Hill Notation):
C15H14N2OS
CAS Number:
Molecular Weight:
270.35

Products

Feature
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade