
Fangchinoline

Synonym(s):
(+)-Limacine, 7-O-Demethyltetrandine, Frangchinoline, Limacine
Empirical Formula (Hill Notation):
C37H40N2O6
CAS Number:
Molecular Weight:
608.72
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing