
EX-CELL® 293 Serum-Free Medium for HEK 293 Cells

Synonym(s):
HEK 293 Medium
Products
Product Category
Brand
Physical Form
Glucose (g/L)
HEPES (mM)
L-Glutamine
Sodium Bicarbonate
Sterility
Sterilization Method
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing