
Esomeprazole sodium

Synonym(s):
5-Methoxy-2-[(S)-[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]-1H-benzimidazole sodium salt
Empirical Formula (Hill Notation):
C17H18NaN3O3S
CAS Number:
Molecular Weight:
367.40
Products
Formula Weight
Manufacturer
Agency
Format
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing