
Erythromycin B

Synonym(s):
12-Deoxyerythromycin
Empirical Formula (Hill Notation):
C37H67NO12
CAS Number:
Molecular Weight:
717.93
Products
Product Category
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing