Skip to Content
BO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
CryoCube
®
F740h
Synonym(s):
Eppendorf
®
CryoCube
®
F740h
Products
Product Category
freezers (12)
Feature
Manufacturer
Capacity (L)
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
EPF740540035
Eppendorf
®
CryoCube
®
F740h,
capacity 740 L, cooling, 5 Classic interface, AC/DC input 120 - 208 V, 60 Hz
Expand
View Pricing
EPF740540015
Eppendorf
®
CryoCube
®
F740h,
capacity 740 L, cooling, 3 classic interface, AC/DC input 120 - 208 V, 60 Hz
Expand
View Pricing
EPF571242035
Eppendorf
®
CryoCube
®
F740h,
capacity 570 L, classic interface, handle left side, 5 shelves, CO2 backup system, AC/DC input 115 V, US 3-pin plug
Expand
View Pricing
EPF740543015
Eppendorf
®
CryoCube
®
F740h,
capacity 740 L, classic interface, handle left side, 3 shelves, LN2 backup system, AC/DC input 120—220 V, US 3-pin plug (NEMA L5-15 twist-lock cable)
Expand
View Pricing
EPF740542015
Eppendorf
®
CryoCube
®
F740h,
capacity 740 L, classic interface, handle left side, 3 shelves, CO2 backup system, AC/DC input 208 V, US 3-pin plug (NEMA L5-15 twist-lock cable)
Expand
View Pricing
EPF740543025
Eppendorf
®
CryoCube
®
F740h,
capacity 740 L, classic interface, handle right side, 3 shelves, AC/DC input 120 - 208 V, 60 Hz (US (NEMA L5-15 twist-lock cable)), LN2 backup system
Expand
View Pricing
EPF740540025
Eppendorf
®
CryoCube
®
F740h,
capacity 740 L, classic interface, handle right side, 3 shelves, AC/DC input 120 - 208 V, US 3-pin plug
Expand
View Pricing
EPF571243035
Eppendorf
®
CryoCube
®
F740h,
capacity 570 L, classic interface, handle left side, 5 shelves, LN2 backup system, AC/DC input 115 V, US 3-pin plug
Expand
View Pricing
EPF740542035
Eppendorf
®
CryoCube
®
F740h,
capacity 740 L, classic interface, VIP, green cooling liquids, air cooling, handle left side, 5 shelves, CO2 backup system, AC/DC input 208 V, 60 Hz, US 3-pin plug (NEMA L5-15 twist-lock cable)
Expand
View Pricing
EPF740542045
Eppendorf
®
CryoCube
®
F740h,
capacity 740 L, classic interface, handle right side, 5 shelves, CO2 backup system, AC/DC input 120 - 208 V, US 3-pin plug (NEMA L5-15 twist-lock cable)
Expand
View Pricing
EPF740543035
Eppendorf
®
CryoCube
®
F740h,
capacity 740 L, classic interface, VIP, green cooling liquids, air cooling, handle left side, 5 shelves, LN2 backup system, AC/DC input 120 - 208 V, 60 Hz (60US (NEMA L5-15 twist-lock cable))
Expand
View Pricing
EPF740542025
Eppendorf
®
CryoCube
®
F740h,
capacity 740 L, classic interface, handle right side, 3 shelves, CO2 backup system, AC/DC input 120 - 208 V, 60 Hz (US (NEMA L5-15 twist-lock cable))
Expand
View Pricing
EPF740543045
Eppendorf
®
CryoCube
®
F740h,
capacity 740 L, classic interface, handle right side, 5 shelves, LN2 backup system, AC/DC input 120 - 208 V, US 3-pin plug (NEMA L5-15 twist-lock cable)
Expand
View Pricing
EPF740540045
Eppendorf
®
CryoCube
®
F740h,
capacity 740 L, classic interface, Vhandle right side, 5 shelves, AC/DC input 120 - 208 V, US 3-pin plug
Expand
View Pricing