
Epoxybergamottin

Synonym(s):
6′,7′-Epoxybergamottin, 4-[[(2E)-5-(3,3-Dimethyl-2-oxiranyl)-3-methyl-2-penten-1-yl]oxy]-7H-furo[3,2-g][1]benzopyran-7-one, 5-(6′,7′-Epoxy)geranyloxypsoralen, Bergamottin 6′,7′-epoxide, Epoxybergamottin
Empirical Formula (Hill Notation):
C21H22O5
CAS Number:
Molecular Weight:
354.40
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing