Skip to Content
Merck
(+)-S-Trityl-L-cysteine 97%

(+)-S-Trityl-L-cysteine

(+)-S-Trityl-L-cysteine 97%
Synonym(s):
S-Trityl-L-cysteine
Linear Formula:
(C6H5)3CSCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
363.47

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing