Skip to Content
Merck
(±)-Methadone-D3 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

(±)-Methadone-D3 solution

(±)-Methadone-D3 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
DL-Methadone-d3 solution, DL-6-Dimethylamino-4,4-diphenyl-3-heptanone-1,1,1-d3 solution, DL-Methadone-(heptyl-1,1,1-d3) solution
Empirical Formula (Hill Notation):
C21D3H24NO
CAS Number:
Molecular Weight:
312.46
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique