Skip to Content
Merck
Dimethyl (1-diazo-2-oxopropyl)phosphonate solution ~10% in acetonitrile (H-NMR), ≥96% (HPLC)

Dimethyl (1-diazo-2-oxopropyl)phosphonate solution

Dimethyl (1-diazo-2-oxopropyl)phosphonate solution ~10% in acetonitrile (H-NMR), ≥96% (HPLC)
Synonym(s):
Bestmann-Ohira Reagent, Dimethyl (1-azoacetonyl)phosphonate solution, Dimethyl (acetyldiazomethyl)phosphonate solution
Empirical Formula (Hill Notation):
C5H9N2O4P
CAS Number:
Molecular Weight:
192.11

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1