
Digoxin

Synonym(s):
12β-Hydroxydigitoxin, Digoxin
Empirical Formula (Hill Notation):
C41H64O14
CAS Number:
Molecular Weight:
780.94
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Purity
Agency
Format
Quality Segment
Functional Group
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique