
Diethylene glycol di(p-toluenesulfonate)

Synonym(s):
2,2′-Oxydiethyl ditosylate, Bis(2-tosyloxyethyl)ether, Diethylene glycol ditosylate
Linear Formula:
(CH3C6H4SO3CH2CH2)2O
CAS Number:
Molecular Weight:
414.49
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing