Decaborane(14)
Synonym(s):
Boron hydride, Decaborane, Decaboron tetradecahydride, NSC 39828, Tetradecahydrodecaborane, nido-Decaborane(14)
Linear Formula:
B10H14
Products
Brand
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
Physical Form
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing