
D-Camphor Impurity J, endo-borneol

Synonym(s):
(−)-Borneol, endo-(1S)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing