
(+)-Biotinamidohexanoic acid hydrazide

Synonym(s):
(+)-Biotin-ε-aminocaproyl hydrazide, (+)-Biotin-X-hydrazide, (+)-Biotinamidocaproyl hydrazide, BACH, N-(+)-Biotinyl-6-aminohexanoic hydrazide
Empirical Formula (Hill Notation):
C16H29N5O3S
CAS Number:
Molecular Weight:
371.50
Products
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing