D-Arabinose-1-13C
Empirical Formula (Hill Notation):
13CC4H10O5
CAS Number:
Molecular Weight:
151.12
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing