
Cyprodinil

Synonym(s):
4-Cyclopropyl-6-methyl-N-phenylpyrimidin-2-amine
Empirical Formula (Hill Notation):
C14H15N3
CAS Number:
Molecular Weight:
225.29
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2