
Cyclophosphamide Related Compound A

Synonym(s):
Bis(2-chloroethyl)amine hydrochloride
Linear Formula:
(ClCH2CH2)2NH·HCl
CAS Number:
Molecular Weight:
178.49
Products
Formula Weight
Manufacturer
Melting Point (°C)
Application
Markush Class
Markush Group
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing