
Clarithromycin Impurity H

Synonym(s):
N-Demethyl-N-formyl-6-O-methylerythromycin, 3"-N-Demethyl-3"-N-formylclarithromycin, N-Formyl clarithromycin
Empirical Formula (Hill Notation):
C38H67NO14
CAS Number:
Molecular Weight:
761.94
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2