Skip to Content
Merck
Chitosan biomedical research grade, high molecular weight

Chitosan

Chitosan biomedical research grade, high molecular weight
Synonym(s):
Deacetylated chitin, Poly(D-glucosamine)
Linear Formula:
(C6H11O4N)n
CAS Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Physical Form
Purity
Tissue
Quality Segment
Application
Polymer Composition
Polymer Type
Special Grade