
Calcitonin Salmon Related Compound B

Synonym(s):
Calcitonin salmon-glycine
Empirical Formula (Hill Notation):
C147H242N44O50S2
CAS Number:
Molecular Weight:
3489.89
Products
Formula Weight
Manufacturer
Physical Form
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing