
Byakangelicin

Synonym(s):
9-(2,3-Dihydroxy-3-methylbutoxy)-4-methoxy-7H-furo[3,2-g][1]benzopyran-7-one
Empirical Formula (Hill Notation):
C17H18O7
CAS Number:
Molecular Weight:
334.32
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing