
Butoconazole nitrate

Synonym(s):
(±)-1-[4-(4-Chlorophenyl)-2-[(2,6-dichlorophenyl)thio]butyl]-1H-imidazole mononitrate
Empirical Formula (Hill Notation):
C19H17Cl3N2S · HNO3
CAS Number:
Molecular Weight:
474.79
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing