
Bromhexine for system suitability

Synonym(s):
Bromhexine hydrochloride, 2-Amino-3,5-dibromo-N-cyclohexyl-N-methylbenzylamine hydrochloride, N-(2-Amino-3,5-dibromobenzyl)-N-methylcyclohexylamine hydrochloride, Bromohexine hydrochloride
Empirical Formula (Hill Notation):
C14H20Br2N2 · HCl
CAS Number:
Molecular Weight:
412.59
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing