Skip to Content
Merck
Biotin 4-amidobenzoic acid sodium salt ≥95%

Biotin 4-amidobenzoic acid sodium salt

Biotin 4-amidobenzoic acid sodium salt ≥95%
Synonym(s):
N-(+)-Biotinyl-4-aminobenzoic acid sodium salt, N-Biotinyl-p-aminobenzoic acid sodium salt, Sodium N-(+)-biotinyl-4-aminobenzoate
Empirical Formula (Hill Notation):
C17H20N3NaO4S
CAS Number:
Molecular Weight:
385.41

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing