
Betamethasone Valerate Related Compound A

Synonym(s):
Betamethasone 21-valerate
Empirical Formula (Hill Notation):
C27H37FO6
CAS Number:
Molecular Weight:
476.58
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2