
Anti-TRIM22 antibody produced in rabbit

Synonym(s):
Anti-50 kDa-stimulated trans-acting factor antibody produced in rabbit, Anti-RING finger protein 94 antibody produced in rabbit, Anti-Staf-50 antibody produced in rabbit, Anti-Tripartite motif-containing protein 22 antibody produced in rabbit, Anti-GPSTAF50, Anti-RNF94, Anti-STAF50, Anti-Tripartite motif-containing 22
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique