
Anti-LAMC1 antibody produced in rabbit

Synonym(s):
Anti-Laminin B2 chain antibody produced in rabbit, Anti-Laminin subunit γ-1 precursor antibody produced in rabbit, LAMB2, Laminin B2 chain, Laminin subunit γ-1
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing