Skip to Content
Merck
ANTI-FLAG® M2 Affinity Gel purified immunoglobulin, buffered aqueous glycerol solution

ANTI-FLAG® M2 Affinity Gel

ANTI-FLAG® M2 Affinity Gel purified immunoglobulin, buffered aqueous glycerol solution
Synonym(s):
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, Anti-ddddk, Anti-dykddddk

Products

Product Category
Brand
Clonality
Isotype
Physical Form
Analyte Suitability
Antibody Form
Conjugate
Quality Segment
Technique
Showing 1-1 of 1