
Amrinone Related Compound C

Synonym(s):
1,6-Dihydro-6-oxo(3,4′-bipyridine)-5-carbonitrile
Empirical Formula (Hill Notation):
C11H7N3O
CAS Number:
Molecular Weight:
197.19
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing