Ammonium dihydrogen phosphate
Synonym(s):
Ammonium dihydrogen phosphate, Ammonium phosphate monobasic, mono-Ammonium phosphate, prim-Ammonium phosphate, Ammonium dihydrogenphosphate
Linear Formula:
NH4H2PO4
CAS Number:
Molecular Weight:
115.03
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing