
Acetophenone-β,β,β-d3

Synonym(s):
Acetophenone-(methyl-d3)
Linear Formula:
C6H5COCD3
CAS Number:
Molecular Weight:
123.17
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing