
7-Hydroxycoumarin-3-carboxylic acid

Synonym(s):
Umbelliferone-3-carboxylic acid
Empirical Formula (Hill Notation):
C10H6O5
CAS Number:
Molecular Weight:
206.15
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
pK Value
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing