Skip to Content
BO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
6-Methyl-4-Phenylchromanone
Synonym(s):
6-Methyl-4-phenyl-2-chromanone
Linear Formula:
C
16
H
14
O
2
CAS Number:
40546-94-9
Products
Manufacturer
Agency
Format
Application
Reference Material Type
Special Grade
pharmaceutical analytical impurity (PAI) (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
1A01060
6-Methyl-4-Phenylchromanone,
Pharmaceutical Analytical Impurity (PAI)
Expand
View Pricing