
4-Nitrophenethylamine hydrochloride

Synonym(s):
2-(4-Nitrophenyl)ethylamine hydrochloride
Linear Formula:
O2NC6H4CH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
202.64
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing