Skip to Content
Merck
4-Bromo-2,6-difluoroaniline 98%

4-Bromo-2,6-difluoroaniline

4-Bromo-2,6-difluoroaniline 98%
Linear Formula:
BrC6H2(F)2NH2
CAS Number:
Molecular Weight:
208.00

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing