Skip to Content
Merck
4arm-PEG20K 3arm-OH, 1arm-NH2 HCl Salt, average Mn 20000

4arm-PEG20K

4arm-PEG20K 3arm-OH, 1arm-NH2 HCl Salt, average Mn 20000
Synonym(s):
4arm-PEG-Maleimide, 4arm-PEG-SH, 4arm-PEG-Acrylate, 4arm-PEG-NH2, 4arm-PEG-Isocyanate, 4arm-PEG-Succinimidyl Carboxymethyl Ester, 4arm-PEG-Vinylsulfone, 4arm-PEG-Succinimidyl Succinate
Linear Formula:
C(CH2O(CH2CH2O)nCH2CH2NH2HCl)(CH2O(CH2CH2O)nH)3