
1-Ethyl-3-methylimidazolium chloride

Synonym(s):
1-Ethyl-3-methylimidazolium chloride
Empirical Formula (Hill Notation):
C6H11ClN2
CAS Number:
Molecular Weight:
146.62
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-5 of 5