
17,21-Dihydroxypregna-1,4,9(11)-triene-3,20-dione

Synonym(s):
Deltacortinene
Empirical Formula (Hill Notation):
C21H26O4
CAS Number:
Molecular Weight:
342.43
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1