Skip to Content
Merck
All Photos(1)

Key Documents

1545056

USP

Plastic additive 14

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Bis(2-ethylhexyl) phthalate, DEHP, Phthalic acid bis(2-ethylhexyl ester), ‘Dioctyl’ phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H38O4
CAS Number:
Molecular Weight:
390.56
Beilstein:
1890696
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

USP

bp

386 °C (lit.)

density

0.985 g/mL at 20 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCC(CC)COC(=O)c1ccccc1C(=O)OCC(CC)CCCC

InChI

1S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20H,5-10,13-14,17-18H2,1-4H3

InChI key

BJQHLKABXJIVAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Plastic additive 14 USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

392.0 °F - Pensky-Martens closed cup

Flash Point(C)

200 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service