Skip to Content
Merck
All Photos(1)

Key Documents

24765

Supelco

Screw cap (open-top), polypropylene

yellow polypropylene hole cap, thread for 8-425, no septum, pkg of 1000 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

material

yellow polypropylene hole cap

feature

autoclavable
closure type black polypropylene hole cap
closure type screw top vial
no septum

packaging

pkg of 1000 ea

parameter

135 °C temp. limit

fitting

thread for 8-425

compatibility

for use with 2 mL vial, 12 × 32 mm, 4.6 mm I.D.

Looking for similar products? Visit Product Comparison Guide

General description

These hole caps do not come with septa.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hadas Cohen-Dvashi et al.
Cell host & microbe, 27(3), 418-427 (2020-02-16)
Ebola virus disease is a severe health problem in Africa. Vaccines that display the Zaire ebolavirus glycoprotein spike complex are a prime component for the effort to combat it. The VH3-15/Vλ1-40-based class of antibodies was recently discovered to be a
Hiroki Saito et al.
Nutrients, 12(6) (2020-06-04)
The beneficial effects of fatty acids (FAs) on human health have attracted widespread interest. However, little is known about the impact of FAs on the handling of urate, the end-product of human purine metabolism, in the body. Increased serum urate
Atilgan Yilmaz et al.
Cell stem cell, 27(4), 679-691 (2020-08-01)
Pluripotent stem cells can differentiate into all embryonic germ layers, yet the genes essential for these cell fate transitions in human remain elusive. Here, we mapped the essential genes for the differentiation of human pluripotent stem cells (hPSCs) into the
Alfredo M Valencia et al.
Cell, 179(6), 1342-1356 (2019-11-25)
Mammalian switch/sucrose non-fermentable (mSWI/SNF) complexes are multi-component machines that remodel chromatin architecture. Dissection of the subunit- and domain-specific contributions to complex activities is needed to advance mechanistic understanding. Here, we examine the molecular, structural, and genome-wide regulatory consequences of recurrent
Yoichiro Fujioka et al.
Cell host & microbe, 23(6), 809-818 (2018-05-22)
Influenza A virus (IAV) infection is initiated by the attachment of the viral glycoprotein hemagglutinin (HA) to sialic acid on the host cell surface. However, the sialic acid-containing receptor crucial for IAV infection has remained unidentified. Here, we show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service