Skip to Content
Merck
All Photos(1)

Key Documents

SRP6311

Sigma-Aldrich

Trypsin Pancreas from human pancreas

≥95% (SDS-PAGE)

Synonym(s):

Beta-trypsin, Cationic trypsinogen, PRSS1, Serine protease 1, TRP1, TRY1, TRYP1, Trypsin I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

biological source

human

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

21-23 kDa

packaging

pkg of 50 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TRYP1(5644)

Related Categories

General description

Trypsin hydrolyzes peptides, amides, and ester bonds involving the carboxyl groups of L-arginine or L-lysine. Increased serum values of this enzyme and/or its zymogen (trypsinogen) have been found in patients with cystic fibrosis. Trypsinogen has three isoforms: cationic (trypsinogen 1), anionic (trypsinogen 2, MW - 22.5 kDa), and mesotrypsinogen (trypsinogen 3, MW - 25 kDa). The trypsin available is the cationic form.

Physical form

Lyophilized from 2 mM HCl.

Reconstitution

In water or aqueous buffer

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service