Skip to Content
Merck
All Photos(1)

Key Documents

SML3293

Sigma-Aldrich

3′,2′-Cgamp Disodium

≥98% (HPLC)

Synonym(s):

Adenylyl-(2′→5′)-3′-guanylic acid cyclic nucleotide disodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N10O13P2 ·2Na
Molecular Weight:
718.38
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

storage temp.

−20°C

SMILES string

NC1=NC=NC2=C1N=CN2[C@H]3[C@H](OP4([O-])=O)[C@H](O)[C@@H](COP([O-])(O[C@H]5[C@@H](O)[C@H](N(C=N6)C7=C6C(NC(N)=N7)=O)O[C@@H]5CO4)=O)O3.[Na].[Na+]

General description

3’,2’-cGAMP is a signaling molecule produced by the host immune system in response to pathogens. 3’,2’-cGAMP is synthesized from CdnG , one of the Cyclic-oligonucleotide-based antiphage signaling systems (CBASS). It activates the stimulator of interferon genes (STING)effector protein to initiate a potent interferon response. The drosophila STING (dSTING) in complex with 3′2′-cGAMP explains selective isomer recognition.

Biochem/physiol Actions

3′,2′-cGAMP is a bacterial second messenger involved in antiphage defense system. 3′,2′-cGAMP is a signaling molecule that activates Cap5 for DNA degradation in Asticcacaulis sp. and Lactococcus lactis. 3′2′-cGAMP is produced upon in Drosophila melanogaster upon activation by viral RNA or DNA of cGAS-like receptors (cGLRs) and induces antiviral response.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shirin Fatma et al.
Nature communications, 12(1), 6381-6381 (2021-11-06)
Cyclic-oligonucleotide-based antiphage signaling systems (CBASS) are diverse and abundant in bacteria. Here, we present the biochemical and structural characterization of two CBASS systems, composed of CdnG and Cap5, from Asticcacaulis sp. and Lactococcus lactis. We show that CdnG from Asticcacaulis
Pivotal Roles of cGAS-cGAMP Signaling in Antiviral Defense and Immune Adjuvant Effects
LI ZG, et al.
Science (2013)
Andreas Holleufer et al.
Nature, 597(7874), 114-118 (2021-07-15)
In mammals, cyclic GMP-AMP (cGAMP) synthase (cGAS) produces the cyclic dinucleotide 2'3'-cGAMP in response to cytosolic DNA and this triggers an antiviral immune response. cGAS belongs to a large family of cGAS/DncV-like nucleotidyltransferases that is present in both prokaryotes1 and
Kailey M Slavik et al.
Nature, 597(7874), 109-113 (2021-07-15)
Cyclic GMP-AMP synthase (cGAS) is a cytosolic DNA sensor that produces the second messenger cG[2'-5']pA[3'-5']p (2'3'-cGAMP) and controls activation of innate immunity in mammalian cells1-5. Animal genomes typically encode multiple proteins with predicted homology to cGAS6-10, but the function of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service