Skip to Content
Merck
All Photos(1)

Key Documents

SML1331

Sigma-Aldrich

MLS1547

≥98% (HPLC)

Synonym(s):

5-Chloro-7-[[4-(2-pyridinyl)-1-piperazinyl]methyl]-8-quinolinol, MLS000051547

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H19ClN4O
CAS Number:
Molecular Weight:
354.83
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

OC1=C(CN2CCN(C3=NC=CC=C3)CC2)C=C(Cl)C4=CC=CN=C41

InChI

1S/C19H19ClN4O/c20-16-12-14(19(25)18-15(16)4-3-7-22-18)13-23-8-10-24(11-9-23)17-5-1-2-6-21-17/h1-7,12,25H,8-11,13H2

InChI key

OPEJNANYABTIGC-UHFFFAOYSA-N

Biochem/physiol Actions

MLS1547 is a highly efficacious D2 dopamine receptor (D2R) G protein-biased agonist that does not recruit β-arrestin, and is an antagonist of dopamine-stimulated β-arrestin recruitment to the D2 receptor. MLS-1547 is the first D2 receptor agonist with close to full efficacy for G protein signaling (EC50 = 370 nM), but no ability to recruit β-arrestin.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Benjamin Free et al.
Molecular pharmacology, 86(1), 96-105 (2014-04-24)
A high-throughput screening campaign was conducted to interrogate a 380,000+ small-molecule library for novel D2 dopamine receptor modulators using a calcium mobilization assay. Active agonist compounds from the primary screen were examined for orthogonal D2 dopamine receptor signaling activities including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service