Skip to Content
Merck
All Photos(5)

Key Documents

G1126

Sigma-Aldrich

DL-Glutamic acid monohydrate

≥98% (HPLC)

Synonym(s):

(±)-2-Aminoglutaric acid, (±)-2-Aminopentanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2CH2CH(NH2)COOH · H2O
CAS Number:
Molecular Weight:
165.14
Beilstein:
4155097
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

DL-Glutamic acid monohydrate, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white

mp

180 °C

solubility

1 M HCl: soluble

SMILES string

O.NC(CCC(O)=O)C(O)=O

InChI

1S/C5H9NO4.H2O/c6-3(5(9)10)1-2-4(7)8;/h3H,1-2,6H2,(H,7,8)(H,9,10);1H2

InChI key

OZDAOHVKBFBBMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

DL-Glutamic acid is a racemic mixture of the proteinogenic amino acid L-Glutamic acid and the non-proteinogenic amino acid D-Glutamic acid. DL-Glutamic acid is used ito study its mechanisms of crystal formation and self-assembly on surfaces and copolymer development.

Biochem/physiol Actions

Excitatory amino acid neurotransmitter that is an agonist at kainate, NMDA, AMPA, and metabotropic glutamatergic receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan Jiang et al.
Physical chemistry chemical physics : PCCP, 14(2), 914-919 (2011-11-29)
The existence region of a polymer-induced liquid precursor (PILP) phase for crystals of an organic compound (DL-glutamic acid, Glu) was determined for the first time in the phase diagram of the Glu-polyethyleneimine-water-ethanol system. The existence region and the amount of
Pradip Paik et al.
ACS applied materials & interfaces, 1(8), 1834-1842 (2010-04-02)
In this work, we synthesized chiral mesoporous silica (CMS) spheres, which can be used as a potential candidate for chiral separation. The CMS spheres with controllable pore sizes (of 2-3 nm) and high surface areas of ca. 614 m(2) g(-1)
Michal Ejgenberg et al.
Chemical communications (Cambridge, England), 47(44), 12161-12163 (2011-10-12)
In this communication, we demonstrate that chiral self-assembled monolayers can be used for polymorphism control of chiral crystals. We studied the crystallization of DL-glutamic acid on chiral self-assembled monolayers and showed that crystallization of DL-glutamic acid on the chiral SAMs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service