Skip to Content
Merck
All Photos(1)

Key Documents

EHU035381

Sigma-Aldrich

MISSION® esiRNA

targeting human SGK1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACCCTTCTCCTCCACCAAGTCCTTCTCAGCAAATCAACCTTGGCCCGTCGTCCAATCCTCATGCTAAACCATCTGACTTTCACTTCTTGAAAGTGATCGGAAAGGGCAGTTTTGGAAAGGTTCTTCTAGCAAGACACAAGGCAGAAGAAGTGTTCTATGCAGTCAAAGTTTTACAGAAGAAAGCAATCCTGAAAAAGAAAGAGGAGAAGCATATTATGTCGGAGCGGAATGTTCTGTTGAAGAATGTGAAGCACCCTTTCCTGGTGGGCCTTCACTTCTCTTTCCAGACTGCTGACAAATTGTACTTTGTCCTAGACTACATTAATGGTGGAGAGTTGTTCTACCATCTCCAGAGGGAACGCTGCTTCCTGGAACCACGGGCTCGTTTCTATGCTGCTGAAATAGCCAGTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadeshda Schelski et al.
Pflugers Archiv : European journal of physiology, 471(6), 889-899 (2019-02-02)
The serum- and glucocorticoid-inducible kinase 1 (SGK1) is a key regulator of osteo-/chondrogenic transdifferentiation and subsequent calcification of vascular smooth muscle cells (VSMCs). The phenotypical transdifferentiation of VSMCs is associated with increased interleukin-18 (IL-18) levels and generalized inflammation. Therefore, the
Florian Poetsch et al.
International journal of molecular sciences, 21(19) (2020-10-03)
In diabetes mellitus, hyperglycemia promotes the osteogenic transdifferentiation of vascular smooth muscle cells (VSMCs) to enhance medial vascular calcification, a common complication strongly associated with cardiovascular disease and mortality. The mechanisms involved are, however, still poorly understood. Therefore, the present
Eneda Toska et al.
Cell reports, 27(1), 294-306 (2019-04-04)
The PI3K pathway integrates extracellular stimuli to phosphorylate effectors such as AKT and serum-and-glucocorticoid-regulated kinase (SGK1). We have previously reported that the PI3K pathway regulates estrogen receptor (ER)-dependent transcription in breast cancer through the phosphorylation of the lysine methyltransferase KMT2D
Jakob Voelkl et al.
The Journal of clinical investigation, 128(7), 3024-3040 (2018-06-12)
Medial vascular calcification, associated with enhanced mortality in chronic kidney disease (CKD), is fostered by osteo-/chondrogenic transdifferentiation of vascular smooth muscle cells (VSMCs). Here, we describe that serum- and glucocorticoid-inducible kinase 1 (SGK1) was upregulated in VSMCs under calcifying conditions.
Ferran Medina-Jover et al.
FEBS letters, 594(19), 3200-3215 (2020-08-09)
BMP9 is a cytokine involved in the maturation phase of the angiogenic process that signals through its serine/threonine receptor ALK1 and its coreceptor endoglin. In this paper, we explain how BMP9 directs the regulation of endothelial cell proliferation blockage while

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service