Skip to Content
Merck
All Photos(1)

Key Documents

SK.N.SH Cell Line human

86012802, human nerve (bone marrow metastasis; Caucasian neuroblastoma), Epithelial

Synonym(s):

NSH Cells, SK-NSH Cells, SKNSH Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

SK.N.SH Cell Line human, 86012802, from human nerve(bone marrow metastasis; Caucasian neuroblastoma)

biological source

human nerve (bone marrow metastasis; Caucasian neuroblastoma)

growth mode

Adherent

karyotype

2n = 46, modal no. 47

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian neuroblastoma

Cell Line Description

Established from a bone marrow metastasis from a 4-year-old caucasian female suffering with neuroblastoma. The cells have been used as a target cell in cell-mediated cytoxicity tests.

Application

Human SK.N.SH cell line has been used in dark toxicity testing of a chloro-indium-phthalocyanine photosensitizer (In-Pc). It has also been used to study the potential mechanisms and domain requirements of the hHSJ1a (DnaJ heat shock protein family member B2) mediated reduction in superoxide dismutase 1 (SOD1) aggregation.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 11
D16S539: 8,13
D5S818: 12
D7S820: 7,10
THO1: 7,10
TPOX: 8,11
vWA: 14,18

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:10 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA, RNA, cDNA derived from ECACC mammalian cell lines allow screening for genes or expression patterns to identify lines most suitable for specific research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service