Skip to Content
Merck
All Photos(1)

Key Documents

AC-0369

Sigma-Aldrich

HMGA2 (EP398) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

Ig fraction of antiserum

clone

monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:60

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

General description

High-mobility group AT-hook 2 (HMGA2) is a chromatin factor that regulates transcription and cellular differentiation during embryonic development. HMGA2 helps to regulate the cell cycle and plays an important role in chromosome condensation during the meiotic G2/M transition of spermatocytes. It also interacts with transcription factor E4F1, protein kinase NEK2, and the promoter of the nucleotide excision-repair gene ERCC1 to negatively modulate its activity. HMGA2 expression in adult tissues is often associated with benign and malignant tumor formation. HMGA2 is expressed in most conventional and intramuscular lipomas, 84 and 100%, respectively and can aid in differentiating between lipomas from dedifferentiated liposarcomas and distinguishing areas of tumor from normal adipose tissue. In mesenchymal tumors, HMGA2 is expressed in 88% of benign fibrous histiocytoma, 90% of nodular fasciitis, and 90% of vulvovaginal angiomyxoma. Increased expression of HMGA2 occurs in other human malignancies, including pancreatic, gastric, oral squamous cell, ovarian, and a subset of breast cancers. In thyroid carcinomas, upregulation of HMGA2 can distinguish between benign and malignant follicular neoplasias. Further, HMGA2 overexpression is often found in non-small cell lung cancer and could be used as a marker for lung carcinomas. The presence of HMGA2 in cancer is correlative with metastasis and poor prognosis.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and &lt0.1% Sodium Azide

Other Notes

For a copy of the IFU and CofA contact IVDorder@ABCAM.com

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service