Skip to Content
Merck
All Photos(1)

Key Documents

314M-1

Sigma-Aldrich

Cytokeratin 14 (LL002) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

LL002, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (314M-14)
vial of 0.5 mL concentrate (314M-15)
bottle of 1.0 mL predilute (314M-17)
vial of 1.0 mL concentrate (314M-16)
bottle of 7.0 mL predilute (314M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG3

control

squamous carcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... KRT14(3861)

General description

Cytokeratin 14 (CK 14) is a 50 kD polypeptide found in basal cells of squamous epithelia, some glandular epithelia, myoepithelium, and mesothelial cells. Anti-CK 14 has been demonstrated to be useful in differentiating squamous cell carcinomas from other epithelial tumors. This antibody has also been useful in separating oncocytic tumors of the kidney from it′s renal cell mimic, as well as in determining metaplastic carcinomas of the breast.

Linkage

Cytokeratin 14 Positive Control Slides, Product No. 314S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diagnostic Immunohistochemistry: Theranostic and Genomic Applications, 166-176 (2002)
P G Chu et al.
Histopathology, 39(5), 455-462 (2001-12-12)
The cytokeratin 14 (CK14) expression in oncocytomas or oncocytic tumours of various tissue origins has not been established. We have studied CK14 expression in 30 cases of oncocytic tumours of various tissue origins and 33 cases of renal cell carcinoma
Jorge S Reis-Filho et al.
Applied immunohistochemistry & molecular morphology : AIMM, 11(1), 1-8 (2003-03-01)
Metaplastic carcinomas of the breast (MCBs) are unusual neoplasms characterized by an admixture of glandular epithelial components, which frequently exhibit features of squamous differentiation, and mesenchymal malignant components. Regardless of the presence of myoepithelial features in MCB, no consensus concerning
P G Chu et al.
Histopathology, 39(1), 9-16 (2001-07-17)
The tissue distribution of cytokeratin 14 (CK14) in epithelial neoplasms is not well defined. We have evaluated 435 cases of epithelial neoplasm of various origins with cytokeratin 14 monoclonal antibody with special attention to possible use in differential diagnosis. Immunohistochemistry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service